missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Methyl-1,2-oxazol-3-yl) prop-2-Enamid, TRC

Product Code. 30180099
Change view
Click to view available options
Menge:
50 mg
250 mg
Unit Size:
250 Milligramm
50 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30180099 250 mg 250 Milligramm
30180315 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180099 Supplier Toronto Research Chemicals Supplier No. TRCM103460250MG
 Eingeschränkter Artikel
Verfügbar ab: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 196403-11-9
Summenformel C7H8N2O2
Molekulargewicht (g/mol) 152.15
IUPAC-Name N-(5-methyl-1,2-oxazol-3-yl)prop-2-Enamid
SMILES Cc1onc(NC(=O)C=C)c1

Specifications

Chemischer Name oder Material N-(5-Methyl-1,2-oxazol-3-yl)prop-2-enamide
Menge 250 mg
InChI-Formel InChI=1S/C7H8N2O2/c1-3-7(10)8-6-4-5(2)11-9-6/h3-4H,1H2,2H3,(H,8,9,10)
Formelmasse 152.0586
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.