Learn More
MYO1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15012-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Motor protein that may participate in process critical to neuronal development and function such as cell migration, neurite outgrowth and vesicular transport.Spezifikation
| MYO1B | |
| Polyclonal | |
| Unconjugated | |
| MYO1B | |
| MMI alpha, MMIa, MYH 1c, MYO1B, Myosin I alpha, myosin IB, myr1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117057, 17912, 4430 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43795, P46735, Q05096 | |
| Myo1b | |
| MYO1B Fusion Protein Ag6895 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |