Learn More
Invitrogen™ Mycoplasma hominis Monoclonal Antibody (6508)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA547569
Beschreibung
Specificity: Specific for M. hominis. No cross reactivity with related organsims. No cross reactivity with M. genitalium, M. pneumoniae and U. urealyticum. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Mycoplasma hominis is a species of bacteria that lacks a cell wall around their cell membranes. Mycoplasma hominis is one of the most frequently isolated mycoplasmas from the human genital tract and has been linked to pelvic inflammatory disease and preterm labor.
Spezifikation
| Mycoplasma hominis | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG2b |
| ELISA, Immunocytochemistry | |
| 6508 | |
| Unconjugated | |
| mycoplasmen | |
| Proprietary sequence. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.