missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mycobacterium tuberculosis CFP-10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5122324
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody recognizes the CFP10 (Rv3874) protein of Mycobacterium tuberculosis and Mycobacterium bovis. Western Blot: reducing conditions.
Tuberculosis is a chronic bacterial infection which remains one of the leading killers of young adults world wide. Mycobacterium tuberculosis enters the alveoli by airborne transmission. They are engulfed by alveolar macrophages but resist destruction. Although the exact mechanism is not known, it is suspected that that Mycrobacterium tuberculosis survives by preventing the fusion of the phagosome and lysosome. As the bacterium accumulates it forms a tubercle, spreads to the lymph nodes and becomes a systemic infection. Although treatments for tuberculosis do exist, new strains of Mycobacterium tuberculosis that appear to have multiple-drug resistance (MDR) pose a considerable threat to public health.
Spezifikation
| Mycobacterium tuberculosis CFP-10 | |
| Polyclonal | |
| Unconjugated | |
| m tuberculosis; M. tuberculosis | |
| CFP10 (Rv3874) of Mycobacterium tuberculosis. | |
| 100 μg | |
| Primary | |
| 4°C, do not freeze | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur