missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MUSK Polyclonal Antibody

Artikelnummer. 11561852
Ansicht ändern
Click to view available options
Menge:
200 μg
Packungsgröße:
200 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
11561852 200 μg 200 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11561852 Lieferant Invitrogen™ Lieferanten-Nr. PA11741
  • ONLINE EXCLUSIVE PRICE
    710.00 CHF
    603.50 CHF / 200 Mikroliter
    Sparen 106.50 CHF
    15% Rabatt
Alle Werbeaktionen anzeigen
 Eingeschränkter Artikel
Lager vorrätig
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

PA1-1741 detects a predominant band at ~98kD. Other lower MW nonspecific bands of unknown identity were also detected and are likely to be proteolytic fragments.

MUSK plays an important and selective role in many aspects of skeletal muscle development and function through the initial step of postsynaptic differentiation and formation of end-plate bands in cooperation with Rapsyn. Communication between cells is often mediated by receptors on the surface of the target cell that are triggered by specific ligands released by other cells. Receptor tyrosine kinase activity is strictly regulated by the binding of the appropriate ligand to the extracellular active site of the receptor. MuSK has been shown to be part of the agrin signaling complex and plays an important role in the development and architectural maintenance of the neuromuscular junction.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen MUSK
Anwendungen Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gen MUSK
Gen-Zugriffsnummer O15146, Q61006, Q62838
Gen-Alias CMS9; FADS; Mdk4; MGC126323; MGC126324; Mlk; muscle associated receptor tyrosine kinase; muscle localized kinase 2; muscle specific kinase (neural fold/somite kinase 1); muscle, skeletal receptor tyrosine protein kinase; Muscle, skeletal receptor tyrosine-protein kinase; muscle, skeletal, receptor tyrosine kinase; muscle-specific kinase receptor; muscle-specific protein kinase secretory isoform; muscle-specific tyrosine kinase receptor MuSK; muscle-specific tyrosine protein kinase receptor; Muscle-specific tyrosine-protein kinase receptor; MuSK; Nsk1; Nsk2; Nsk3; RP23-170A3.1
Gensymbole MUSK
Wirtsspezies Rabbit
Immunogen Recombinant protein containing residues 210-304 from the extracellular domain of rat MuSK.
Reinigungsverfahren Protein G
Menge 200 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 18198, 4593, 81725
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.