missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscle Phosphofructokinase/PFKM/PFK-1 Antibody (2F11), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-22615MFV500
Additional Details : Gewicht : 0.00970kg
Beschreibung
Muscle Phosphofructokinase/PFKM/PFK-1 Monoclonal antibody specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human samples. It is validated for Western Blot, ELISASpezifikation
Muscle Phosphofructokinase/PFKM/PFK-1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
2F11 | |
Western Blot, ELISA | |
6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase | |
Recombinant human Muscle Phosphofructokinase/PFKM/PFK-1(1-780aa) purified from E. coli | |
0.1 mL | |
Lipid and Metabolism | |
5213 | |
Store at 4C in the dark. | |
IgG2a κ |