missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUM1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12682-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Mutated melanoma-associated antigen 1 (MUM1) is a 78 kDa protein encoded by the MUM1 gene. MUM1 is expressed in a subset of B cells in the light zone of the germinal center, plasma cells, activated T cells and a wide spectrum of related hematolymphoid neoplasms. Antibodies to MUM1 are useful in the sub classification of lymphoid maligcies.Spezifikation
MUM1 | |
Polyclonal | |
Unconjugated | |
MUM1 | |
EXPAND1, HSPC211, MUM 1, MUM1, Protein expandere | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84939 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q2TAK8 | |
MUM1 | |
MUM1 Fusion Protein Ag3371 | |
150 μL | |
Primary | |
Human | |
Liquid |