Learn More
Heidolph™ Multi Reax
Artikelnummer. 17323480 Alle ansehen Heidolph Instruments Produkte
Ansicht ändern
Click to view available options
Steckertyp:
CH-Stecker
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Steckertyp Packungsgröße
17323480 CH-Stecker 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17323480 Lieferant Heidolph™ Lieferanten-Nr. 5451000003
Verfügbar ab: 14-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Multi Reax is a versatile model with a 3 mm shaking orbit.
  • It can accommodate between 12 (16 to 32 mm diameter) and 26 (10 to 16 mm diameter) tubes
  • Reaction vessels with snap-on lids are also safely fixed
  • The 3 mm shaking orbit and a speed control between 150 and 2000 rpm guarantee excellent mixing results
  • The integrated timer (up to 999 minutes) enables the automatic termination of the shaking function
  • After the time has expired, a signal tone is emitted
TRUSTED_SUSTAINABILITY
Spezifikation
Spannung 230 V
Steckertyp CH-Stecker