Learn More
MUC5B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 28118-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The MUC5B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MUC5B. This antibody recognizes human antigen. The MUC5B antibody has been validated for the following applications: IHC, ELISA analysis.
Spezifikation
| MUC5B | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 727897 | |
| IgG |
| Immunohistochemistry, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human |