missing translation for 'onlineSavingsMsg'
Learn More

MTX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 11610-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838803

  • 131.19 CHF / 20 Mikroliter

Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The protein encoded by this gene is highly similar to the metaxin 2 protein from mouse, which has been shown to interact with the mitochondrial membrane protein metaxin 1. Because of this similarity, it is thought that the encoded protein is peripherally associated with the cytosolic face of the outer mitochondrial membrane, and that it is involved in the import of proteins into the mitochondrion. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 7.
Spezifikation

Spezifikation

MTX2
Polyclonal
Unconjugated
MTX2
metaxin 2, MTX2
Rabbit
Antigen Affinity Chromatography
RUO
10651, 288150, 53375
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75431, O88441
Mtx2
MTX2 Fusion Protein Ag2181
20 μL
Primary
Human, Mouse, Rat
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote