missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTHFS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13114-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Contributes to tetrahydrofolate metabolism. Helps regulate carbon flow through the folate-dependent one-carbon metabolic network that supplies carbon for the biosynthesis of purines, thymidine and amino acids.Spezifikation
MTHFS | |
Polyclonal | |
Unconjugated | |
MTHFS | |
HsT19268, Methenyl THF synthetase, MTHFS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10588, 107885, 300886 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49914, Q9D110 | |
MTHFS | |
MTHFS Fusion Protein Ag3781 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |