missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTAP Mouse anti-Human, Mouse, Rat, Clone: 2B1G6, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66706-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes an enzyme that plays a major role in polyamine metabolism and is important for the salvage of both adenine and methionine. The encoded enzyme is deficient in many cancers because this gene and the tumor suppressor p16 gene are co-deleted. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length natures remain unknown.Spezifikation
MTAP | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13126, Q9CQ65 | |
MTAP | |
MTAP Fusion Protein Ag17954 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2B1G6 | |
Unconjugated | |
MTAP | |
c86fus, MSAP, MTA phosphorylase, MTAP, MTAPase | |
Mouse | |
Protein G | |
RUO | |
298227, 4507, 66902 | |
Antibody | |
IgG1 |