missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18339-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the EcoL15 ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 15q.Spezifikation
MRPL15 | |
Polyclonal | |
Unconjugated | |
MRPL15 | |
HSPC145, L15mt, MRP L15, MRP L7, MRPL15, RPML7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27395, 29088, 297799 | |
Antibody | |
IgG |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CPR5, Q9P015 | |
Mrpl15 | |
MRPL15 Fusion Protein Ag13087 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Liquid |