missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRGPRF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00105W
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
May bind to a neuropeptide and may regulate nociceptor function and/or development, including the sensation or modulation of pain. Expressed in the Gut, vas deferens, uterus and aorta; barely detectable in liver, kidney, lung, and salivary gland. In the brain, markedly abundant in the cerebellum.
Spezifikation
| MRGPRF | |
| Polyclonal | |
| MRGPRF | |
| Mas-related G-protein coupled receptor, member F, Mas-related gene F protein, G-protein coupled receptor RTA, Mrgf, Rta, GPR140, GPR168, MGC21621, G-p | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Q96AM1, Q8VCJ6, P23749 | |
| MRGPRF | |
| A synthetic peptide from the 3d cytoplasmic loop of human Mas-related G-protein coupled receptor member F | |
| RUO | |
| 116535, 211577, 266762 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur