Learn More
MPZL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 11787-1-AP-20UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16839093
  • 136.48 CHF / 20 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
This antibody can be used to detect endogenous MPZL2, however, the reason for the additional band of 32 kDa is still unknown.
Epithelial V-like antigen (EVA) is expressed in thymus epithelium and strongly downregulated by thymocyte developmental progression. This gene is expressed in the thymus and in several epithelial structures early in embryogenesis.
Spezifikation
Spezifikation
MPZL2
Polyclonal
Unconjugated
MPZL2
Epithelial V like antigen 1, EVA, EVA1, MPZL2, myelin protein zero like 2
Rabbit
Antigen Affinity Chromatography
RUO
10205, 14012, 300679
-20°C
Liquid
Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60487, O70255
MPZL2
MPZL2 Fusion Protein Ag2419
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen
Berichtigung von Produktinhalten
Name des Produkts
MPZL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated