Learn More
R&D Systems™ Mouse XL GM-CSF, Luminex™ Performance Assay
Click to view available options
Menge:
1 Kit
Packungsgröße:
1 Set
Spezifikation
Spezifikation
| Menge | 1 Kit |
| Spezies | Mouse |
| Gen-ID (Entrez) | 1437 |
| Synonym | colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim |
| Inhalt und Lagerung | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |