missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VE-Statin Isoform 1 Antibody, R&D Systems™

Goat Polyclonal Antibody
245.00 CHF - 612.00 CHF
Spezifikation
| Antigen | VE-Statin/EGFL7 |
|---|---|
| Konzentration | LYOPH |
| Verdünnung | Western Blot 0.1 ug/mL |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18757037
![]() |
R&D Systems
AF3089-SP |
25 μg |
245.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18013042
|
R&D Systems
AF3089 |
100 μg |
612.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
VE-Statin/EGFL7 Polyclonal specifically detects VE-Statin/EGFL7 in Mouse samples. It is validated for Western Blot.Spezifikation
| VE-Statin/EGFL7 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| EGFL7, EGF-like protein 7, EGF-like-domain, multiple 7, epidermal growth factor-like protein 7, MEGF7, MGC111117, Multiple EGF-like domains protein 7, Multiple epidermal growth factor-like domains protein 7, NEU1 protein, NOTCH4-like protein, RP11-251M1.2, Vascular endothelial statin, VEStatin, VE-STATIN, Zneu1 | |
| EGFL7 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9QXT5 | |
| 51162 | |
| E. coli-derived recombinant mouse VE-Statin Isoform 1 Thr21-Leu275 Accession # Q9QXT5 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse VE-Statin Isoform 1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human VE-Statin is observed. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts