missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse TREM-2 His-tag, Recombinant Protein

HEK293 cells Recombinant Protein

Marke:  Invitrogen™ A42621

Additional Details : Gewicht : 0.01060kg

Artikelnummer. 17154283

  • 79.75 CHF / 5 Mikrogramm
Verfügbar ab: 02-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

TREM2 is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL).
Spezifikation

Spezifikation

Q99NH8
PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4
Mouse TREM-2 His-tag
HEK293 cells
RUO
OTTHUMP00000039771; soluble TREM 2; soluble TREM2; sTREM 2; sTREM2; TREM; TREM2; TREM-2; Trem2a; Trem2b; Trem2c; triggering receptor expressed on monocytes 2; Triggering receptor expressed on myeloid cells; Triggering receptor expressed on myeloid cells 2; triggering receptor expressed on myeloid cells 2 A; triggering receptor expressed on myeloid cells 2 b; triggering receptor expressed on myeloid cells 2 c
Mouse
Recombinant
≥ 95%
Control, ELISA, Immunohistochemistry, Western Blot
83433
5 μg
-20°C
<1 EU/μg
Trem2
Unconjugated
Lyophilized
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote