Learn More
Mouse stomach normal tissue lysate - total protein, 1 mg
Product Code. 32152897 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32152897 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32152897 Supplier Abcam Supplier No. ab292561mg
 Restricted Item
Verfügbar ab: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Mouse stomach normal cell/tissue lysate.
  • Species: Mouse
  • Tissue: stomach normal
  • Form: Liquid
Specifications
For Use With (Application) Western Blot