Learn More
R&D Systems™ Mouse SNAP23 (NP_033248.1) VersaClone cDNA
Spezifikation
Spezifikation
| Gen | SNAP23 |
| Zugriffsnummer | NP_033248.1 |
| Gen-ID (Entrez) | 8773 |
| Gen-Alias | HsT17016, SNAP-23, SNAP23A, SNAP23B, synaptosomal-associated protein 23, synaptosomal-associated protein, 23 kD, synaptosomal-associated protein, 23 kDa, Vesicle-membrane fusion protein SNAP-23 |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Spezies | Mouse |
| Produktlinie | R&D Systems |
| Menge | 10 μg |