missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Protein C ELISA Kit

Artikelnummer. 31369804
Ansicht ändern
Click to view available options
Menge:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
31369804 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 31369804 Lieferant Invitrogen™ Lieferanten-Nr. EM522RB
 Eingeschränkter Artikel
Verfügbar ab: 13-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

ELISA

Mouse Protein C ELISA Kit is an enzyme-linked immunosorbent assay for quantitative detection of mouse Coagulation Factor XIV (Protein C) in serum, plasma, and cell culture supernatants.

Protein C is a vitamin K-dependent plasma glycoprotein that is cleaved to its activated form by the thrombin-thrombomodulin complex. Protein C in its activated form contains a serine protease domain and functions in degradation of the activated forms of coagulation factors V and VIII. Protein C is a vitamin K-dependent serine protease produced in the liver and made up of 2 polypeptide chains. The 62 kDa proenzyme is activated by thrombin bound to an endothelial surface receptor and the active enzyme cleaves factor Va and VIIIa and thus inhibits blood coagulation. The molecular weight of the active enzyme is 55 kDa and the normal concentrations in human plasma is approximately 1-3 ng/mL because of the very fast turnover, the proenzyme concentration is approximately 3 μg/mL. Mutations in the Protein C gene have been associated with thrombophilia due to protein C deficiency, neonatal purpura fulminans, and recurrent venous thrombosis.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P33587
Assay-Bereich 9.5-3000 pg/mL
Assay-Empfindlichkeit: 9.5 pg/mL
Konjugat HRP
Immunassay-Kit-Format Sandwich ELISA Kit
Produkttyp ELISA
Probentyp Plasma, Serum, Supernatant
Zielspezies Mouse
Zur Verwendung mit (Geräte) Absorbance Microplate Reader
Gen-Alias Activation peptide; Anticoagulant protein C; APC; autoprothrombin IIA; Blood coagulation factor XIV; EDQVDPRLIDGK; inactivator of coagulation factors Va, VIII; PC; prepro-protein C; Proc; PROC1; protein C; protein C (inactivator of coagulation factors Va and VIIIa); protein C prepropeptide; protein C, inactivator of coagulation factors Va and VIIIa; THPH3; THPH4; type I protein C; vitamin K-dependent protein C; Vitamin K-dependent protein C heavy chain; Vitamin K-dependent protein C light chainActivation peptide; Anticoagulant protein C; APC; autoprothrombin IIA; Blood coagulation factor XIV; EDQVDPRLIDGK; inactivator of coagulation factors Va, VIII; PC; prepro-protein C; Proc; PROC1; protein C; protein C (inactivator of coagulation factors Va and VIIIa); protein C prepropeptide; protein C, inactivator of coagulation factors Va and VIIIa; THPH3; THPH4; type I protein C; vitamin K-dependent protein C; Vitamin K-dependent protein C heavy chain; Vitamin K-dependent protein C light chain;
Gen-ID (Entrez) 19123
Gensymbol PROC
Interassay-VK 12%
Intraassay-VK 10%
Kitinhalt Pre-coated microwell plate, Standard, Biotinylated Detection Antibody, Streptavidin-HRP, Sample Diluent, Wash Buffer Concentrate, Assay Buffer concentrate, Stop Solution, Substrate Solution, Adhesive Plate Covers
Marker oder Farbstoff Biotin
Menge 96 Tests
Kennzeichnung RUO
Versandbedingung Wet Ice
Lagerungsbedingungen -20°C
Ziel Protein C
Prüfzeit 1 hr 20 min
Gesamte Assay-Zeit 4 hr 45 min
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.