missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Pro-MMP-7 Antibody, R&D Systems™

Rat Monoclonal Antibody
Marke: R&D Systems MAB29671-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MMP-7 Monoclonal specifically detects MMP-7 in Mouse samples. It is validated for Western Blot.
Spezifikation
| MMP-7 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| AAA99983 | |
| MMP7 | |
| Mouse myeloma cell line NS0-derived recombinant mouse MMP-7 Leu18-Leu264 Accession # AAA99983 | |
| 25 μg | |
| Primary | |
| Detects the pro form of mouse MMP-7 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse MMP-2, -3, -8, -9, -12, or -24 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 377313 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24, EC 3.4.24.23, matrilysin, Matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4316 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur