missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse pIgR (NP_035212) VersaClone cDNA

Artikelnummer. 18726899
Ansicht ändern
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18726899 10 μg 10 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18726899 Lieferant R&D Systems™ Lieferanten-Nr. RDC2505

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.

  • The Mouse pIgR (NP_035212) VersaClone cDNA clone contains the complete ORF for pIgR, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The pIgR insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY

Specifications

Gen mpIgR
Zugriffsnummer NP_035212
Konzentration 10 μg at 0.2 μg/μL
Zusammensetzung cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gen-ID (Entrez) 5284
Inhalt und Lagerung Store the unopened product at -20 to -80°C
Spezies Mouse
Menge 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.