Learn More
ACROBiosystems Mouse PD-L1 / B7-H1 Protein, His Tag
390.91 CHF - 2522.00 CHF
Spezifikation
Zugriffsnummer | NP_068693 |
---|---|
Molekulargewicht | 26.6 kDa |
Quelle | Mouse PD-L1, His Tag (PD1-M5220) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession # NP_068693). |
Immunogen | Phe 19 - Thr 238 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15807487
|
ACROBiosystems
PD1-M5220-100UG |
100 μg |
390.91 CHF
100 Mikrogramm |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15817487
|
ACROBiosystems
PD1-M5220-1MG |
1 mg |
2522.00 CHF
1 Milligramm |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Programmed cell death 1 ligand 1 (PD-L1) is also known as cluster of differentiation (CD274) or B7 homolog 1 (B7-H1), is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses. B7-H1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. PD-L1 is highly expressed in the heart, skeletal muscle, placenta and lung and weakly expressed in the thymus, spleen, kidney and liver. PD-L1 is expressed on activated T-cells, B-cells, dendritic cells, keratinocytes and monocytes. PD-L1 is up-regulated on T- and B-cells, dendritic cells, keratinocytes and monocytes after LPS and IFNG activation and up-regulated in B-cells activated by surface Ig cross-linking. PD-L1 involve in the costimulatory signal, essential for T-cell proliferation and production of IL10 and IFNG, in an IL2-dependent and a PDCD1-independent manner.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.6 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_068693 | |
Mouse PD-L1, His Tag (PD1-M5220) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession # NP_068693). | |
-20°C | |
PD-L1 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
26.6 kDa | |
Phe 19 - Thr 238 | |
PD-L1,CD274,B7-H1,PDCD1L1,PDCD1LG1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |