Learn More
ACROBiosystems Mouse PD-1 / PDCD1 Protein, His Tag
Marke: ACROBiosystems PD1-M5228-100UG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Programmed cell death protein 1 (PD-1) is also known as CD279 and PDCD1, is a type I membrane protein and is a member of the extended CD28/CTLA-4 family of T cell regulators. PDCD1 is expressed on the surface of activated T cells, B cells, macrophages, myeloid cells and a subset of thymocytes. PD-1 has two ligands, PD-L1 and PD-L2, which are members of the B7 family. PD-L1 is expressed on almost all murine tumor cell lines, including PA1 myeloma, P815 mastocytoma, and B16 melanoma upon treatment with IFN-γ. PD-L2 expression is more restricted and is expressed mainly by DCs and a few tumor lines. PD1 inhibits the T-cell proliferation and production of related cytokines including IL-1, IL-4, IL-10 and IFN-γ by suppressing the activation and transduction of PI3K/AKT pathway. In addition, coligation of PD1 inhibits BCR-mediating signal by dephosphorylating key signal transducer. In vitro, treatment of anti-CD3 stimulated T cells with PD-L1-Ig results in reduced T cell proliferation and IFN-γ secretion. Monoclonal antibodies targeting PD-1 that boost the immune system are being developed for the treatment of cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.0 kDa. The protein migrates as 25-45 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.Spezifikation
NP_032824 | |
100 μg | |
Leu 25 - Gln 167 | |
PDCD1,PD1,CD279,SLEB2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
17.0 kDa | |
Mouse PD-1, His Tag (PD1-M5228) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # NP_032824). | |
-20°C | |
PD-1 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |