missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MCAM/CD146 Antibody, R&D Systems™

Rat Monoclonal Antibody
Marke: R&D Systems MAB7718-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Spezifikation
| MCAM/CD146 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Q8R2Y2 | |
| MCAM | |
| Mouse myeloma cell line NS0-derived recombinant mouse MCAM/CD146 Met1-Val563 Accession # Q8R2Y2 | |
| 25 μg | |
| Primary | |
| Detects mouse MCAM/CD146 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) MAdCAM-1 is observed, and no cross-reactivity with rmALCAM, rmNCAM, rmL1CAM, rmOCAM, rmTROP-2, recombinant human MCAM, or recombinant rat MCAM is observed. | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 733216 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, L-Gicerin, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18, MUC18Gicerin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4162 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido