Learn More
R&D Systems™ Mouse IL1RAPL1 (P59823) VersaClone cDNA
Artikelnummer. 18702139
Ansicht ändern
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18702139 10 μg 10 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18702139 Lieferant R&D Systems™ Lieferanten-Nr. RDC2457
  • ONLINE EXCLUSIVE PRICE
    807.00 CHF
    763.35 CHF / 10 Mikrogramm
    Sparen 43.65 CHF
    5% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
  • The Mouse IL-1 RAPL1/IL-1 R8 (P59823) VersaClone cDNA clone contains the complete ORF for IL-1 RAPL1/IL-1 R8, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The IL-1 RAPL1/IL-1 R8 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY
Spezifikation
Gen mIL-1RAPL1/IL-1R8
Zugriffsnummer P59823
Konzentration 10 μg at 0.2 μg/μL
Zusammensetzung cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gen-ID (Entrez) 11141
Gen-Alias IL1R8IL1RAPL, IL-1R9, IL-1 RAPL1, IL-1-RAPL-1, IL-1 RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MR x 10, MR x 21, MR x 34, oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2, TIGIRR-2 mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1
Inhalt und Lagerung Store the unopened product at -20 to -80°C
Spezies Mouse
Produktlinie R&D Systems
Menge 10 μg