missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse IL-4 R alpha / CD124 Protein, His Tag

Marke:  ACROBiosystems ILR-M52H1-100UG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15817137

  • 299.38 CHF / 100 Mikrogramm


Verfügbar ab
04-06-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

IL-4 is a pleiotropic cytokine produced by activated Th2 cells and mast cells, and plays a pivotal role in immune responses. The effects of IL-4 are mediated after binding to high affinity receptor complexes present on hematopoietic as well as non-hematopoietic cells. Hematopoietic cellular responses to IL-4 are mediated by a high affinity receptor complex comprised of the 140 kDa IL4Rα (CD124)subunit and the 70 kDa common cytokine γc chain (CD132). Interleukin 4 Receptor (IL4R) also known as CD124, IL4Rα and BSF receptor, is a type I cytokine receptor produced by activated Th2 cells and mast cells, and plays an important role in Th2-­biased immune responses, alternative macrophage activation, mucosal immunity, allergic inflammation, tumor progression, and atherogenesis. A soluble form of the encoded IL4R protein can be produced by an alternate splice variant or by proteolysis of the membrane-bound protein, and this soluble form can inhibit IL4-mediated cell proliferation and IL5 upregulation by T-cells. IL4R can alternatively associate with IL­13Ra1 to form the type II receptor which is responsive to both IL­4 and IL­13. Interleukin-4 receptor has been shown to interact with SHC1.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.3 kDa. The protein migrates as 37-45 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation

Spezifikation

NP_001008700
HEK293
Mouse
26.3 kDa
100 µg
Ile 26 - Arg 233
IL4R,CD124,IL4RA
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Mouse IL-4 R alpha, His Tag (ILR-M52H1) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Arg 233 (Accession # NP_001008700).
-20°C
IL-4 R alpha
His Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation