missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-16 Antibody, R&D Systems™

Goat Polyclonal Antibody
236.00 CHF - 562.00 CHF
Spezifikation
| Antigen | IL-16 |
|---|---|
| Konzentration | LYOPH |
| Verdünnung | Western Blot 0.1 ug/mL |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18738627
![]() |
R&D Systems
AF1727-SP |
25 μg |
236.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
18089971
![]() |
R&D Systems
AF1727 |
100 μg |
562.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
IL-16 Polyclonal specifically detects IL-16 in Mouse samples. It is validated for Western Blot.Spezifikation
| IL-16 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| FLJ16806, FLJ42735, FLJ44234, HsT19289, IL16, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCF, LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16 | |
| IL16 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| O54824 | |
| 3603 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-16, Ser1205-Ser1322, Accession # O54824 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse IL-16 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human IL-16 is observed. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts