Learn More
Mouse IgG2b kappa Isotype Control (eBMG2b), Super Bright™ 780, eBioscience™, Invitrogen™
Mouse Isotype Control
156.00 CHF - 367.00 CHF
Spezifikation
Antigen | Mouse IgG2b kappa |
---|---|
Konjugat | Super Bright 780 |
Form | Liquid |
Wirtsspezies | Mouse |
Regulatorischer Status | RUO |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15984174
|
Affymetrix eBioscience
78-4732-82 |
100 μg |
367.00 CHF
100 Mikrogramm |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15974174
|
Invitrogen
78-4732-80 |
25 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin. Applications Reported: This mouse IgG2b isotype control has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis. Applications Tested: Super Bright 780 Mouse IgG2b Isotype Control has been tested by flow cytometric analysis and can be used at the same concentration as the experimental antibody. Super Bright 780 is a tandem dye that can be excited with the violet laser line (405nm) and emits at 780nm. We recommend using a 780/60 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product # SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.
Mouse IgG2b kappa Control antibody specifically detects Mouse IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Mouse IgG2b kappa | |
Liquid | |
RUO | |
Isotype Control (Primary) |
Super Bright 780 | |
Mouse | |
IgG2b κ | |
Affinity chromatography |