Learn More
Mouse IgG2a kappa Isotype Control (eBM2a), Super Bright™ 645, eBioscience™, Invitrogen™
Mouse Isotype Control
189.00 CHF - 439.00 CHF
Spezifikation
Antigen | Mouse IgG2a kappa |
---|---|
Klon | eBM2a |
Konjugat | Super Bright 645 |
Form | Liquid |
Wirtsspezies | Mouse |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15815693
|
Affymetrix eBioscience
64-4724-82 |
100 μg |
439.00 CHF
100 Mikrogramm |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15805693
|
Affymetrix eBioscience
64-4724-80 |
25 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Mouse IgG2a kappa Control antibody specifically detects Mouse IgG2a kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Mouse IgG2a kappa | |
Super Bright 645 | |
Mouse | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Isotype Control (Primary) |
eBM2a | |
Liquid | |
RUO | |
IgG2a κ | |
Affinity chromatography |
For Research Use Only