missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), Super Bright™ 702, eBioscience™, Invitrogen™

Mouse Isotype Control

187.00 CHF - 440.00 CHF

Spezifikation

Antigen Mouse IgG1 kappa
Klon P3.6.2.8.1
Konjugat Super Bright 702
Form Liquid
Wirtsspezies Mouse
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15835743
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
67-4714-82
100 μg
440.00 CHF
100 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15825743
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
67-4714-80
25 μg Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
N/A Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Beschreibung

Beschreibung

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Mouse IgG1 kappa Control antibody specifically detects Mouse IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.
Spezifikation

Spezifikation

Mouse IgG1 kappa
Super Bright 702
Mouse
PBS with BSA and 0.09% sodium azide; pH 7.2
Isotype Control (Primary)
P3.6.2.8.1
Liquid
RUO
IgG1 κ
Affinity chromatography
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

For Research Use Only