Learn More
Gibco™ Mouse IFN-gamma Recombinant Protein, PeproTech®
Beschreibung
Recombinant Murine IFN-gamma is a 134 amino-acid length protein with a predicted molecular weight of 15.6 kDa. C-terminal truncation due to proteolytic cleavage occurs during E. coli fermentation and results in a 15.1 kDa protein confirmed by mass spectroscopy. SDS-PAGE analysis of Recombinant Murine IFN-gamma shows a band that migrates at approximately 11-12 kDa, under non-reducing and reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Spezifikation
Spezifikation
Zugriffsnummer | P01580 |
Zur Verwendung mit (Anwendung) | ELISA, Functional Assay, Western Blot |
Zusammensetzung | protein with no preservative |
Gen-ID (Entrez) | 15978 |
Molekulargewicht | 15.6 kDa |
Name | Mouse IFN-gamma |
Menge | 500 μg |
Quelle | E. coli |
Kennzeichnung | RUO |
Endotoxin-Konzentration | <1 EU/ μg |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.