Learn More
R&D Systems™ Mouse IFN-beta Quantikine ELISA Kit
Designed to measure mouse IFN-beta in cell culture supernates, serum, and plasma.
Marke:  R&D Systems™ MIFNB0
Artikelnummer. 30001926
  • 848.00 CHF / 96 Tests
Nur noch null auf Lager
Lager vorrätig
Beschreibung
The Mouse IFN-beta Quantikine ELISA Kit quantifies mouse IFN-beta in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
15.60 - 1000pg/mL
Mouse IFN-beta Quantikine ELISA Kit
Natural and Recombinant Mouse IFN-beta
3456
1 Kit
Store the unopened product at 2° to 8°C. Do not use past expiration date.
IFN-beta
1.89pg/mL
Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
ELISA
Maus
Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
4.5 hr.
Zertifikate
Aktionen
Aktionen
Berichtigung von Produktinhalten
Name des Produkts
R&D Systems™ Mouse IFN-beta Quantikine ELISA Kit > Mouse IFN-beta Quantikine ELISA Kit
For Research Use Only. Not for use in Humans.