Learn More
Mouse IFN-beta Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Marke: R&D Systems MAB8234-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IFN-beta Monoclonal specifically detects IFN-beta in Mouse samples. It is validated for Neutralization.
Spezifikation
| IFN-beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| Human embryonic kidney cell line HEK293-derived mouse IFN-beta Ile22-Asn182 Accession # P01575 | |
| 100 μg | |
| Primary | |
| Detects mouse IFN-beta in direct ELISAs. | |
| Mouse | |
| IgG |
| Neutralization | |
| 1176D | |
| Neutralization 0.25-1.25 ug/mL | |
| P01575 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3456 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |