Learn More
R&D Systems™ Mouse Erythroferrone/FAM132B (NP_775571) VersaClone cDNA
Spezifikation
Spezifikation
| Gen | FAM132B |
| Zugriffsnummer | NP_775571 |
| Gen-ID (Entrez) | 151176 |
| Gen-Alias | C1qTNF15, Complement C1q Tumor Necrosis Factor-Related Protein 15, CTRP15, ERFE, Erythroferrone, FAM132B, Family With Sequence Similarity 132, Member B, Myonectin, Protein FAM132B |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Spezies | Mouse |
| Menge | 10 μg |