missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse D-Dimer ELISA Kit
Click to view available options
Menge:
96 Tests
Packungsgröße:
96 Stück
Description
Fibrinogen is the main protein of blood coagulation system. It consists of two identical subunits that contain three polypeptide chains: alpha, beta and gamma. The process of blood coagulation results in the activation of fibrinogen into fibrin by thrombin and fibrin polymerization. Fibrin clot is then digested by plasmin, and fibrin degradation products of different molecular weights are released into the bloodstream.
Specifications
Specifications
| Zugriffsnummer | E9PV24 |
| Assay-Bereich | 78,13 bis 5000 ng |
| Assay-Empfindlichkeit: | 46.88 ng/mL |
| Konjugat | HRP |
| Produkttyp | ELISA |
| Probentyp | Plasma |
| Zur Verwendung mit (Geräte) | Kolorimetrisches Mikrotiterplatten-Lesegerät |
| Gen-ID (Entrez) | 14161 |
| Gensymbol | FGA |
| Interassay-VK | <10% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction