missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cripto Antibody, R&D Systems™

Rat Monoclonal Antibody
Marke: R&D Systems MAB1538
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CRIPTO Monoclonal specifically detects CRIPTO in Mouse samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready.
Spezifikation
| CRIPTO | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| P51865 | |
| Tdgf1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cripto Arg26-Gln150 Accession # P51865 | |
| 150 μg | |
| Primary | |
| Detects mouse Cripto in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ART, recombinant mouse (rm) ART, rmArtemin, recombinant drosophila DPP, rhGDNF, recombinant rat (rr) GDNF, rhLAP, rhLatent TGF-beta 1, rhTGF-beta 2, rhLefty-A, rmLefty-1, rrMIS, rhNeurturin, rmNodal, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 3, or recombinant amphibian TGF-beta 5 is observed. | |
| Mouse | |
| Purified |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF | |
| 237603 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CFC-2, CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, TDGF1, TDGF-1, teratocarcinoma-derived growth factor 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6997 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur