missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD300b/LMIR5 Antibody, R&D Systems™

Rat Monoclonal Antibody
Marke: R&D Systems MAB2580-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD300b/LMIR5/CD300LB Monoclonal specifically detects CD300b/LMIR5/CD300LB in Mouse samples. It is validated for Western Blot.
Spezifikation
| CD300b/LMIR5/CD300LB | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q3U497 | |
| CD300LB | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD300b/LMIR5 Ile18-Tyr157 Accession # Q3U497 | |
| 25 μg | |
| Primary | |
| Detects mouse CD300b/LMIR5 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) LMIR5 is observed and no cross-reactivity with recombinant mouse (rm) LMIR3, rmLMIR4, rhLMIR1, rhLMIR2, or rhLMIR6 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 339003 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD300 antigen like family member B, CD300 antigen-like family member B, CD300 molecule-like family member b, CD300B, CD300b antigen, CD300LB, CLM7, CLM-7, CLM7IREM3CD300 molecule like family member b, CMRF35A2, CMRF35-A2, CMRF35-like molecule 7, EC 1.13.11.6, EC 6.3.4.3, Gm253, Immune receptor expressed on myeloid cells 3, IREM-3, Leukocyte mono-Ig-like receptor 5, LMIR5, TREM-5, TREM5CD300b, Triggering receptor expressed on myeloid cells 5 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 124599 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur