missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse CD28 Protein, His Tag

387.68 CHF - 2626.22 CHF

Spezifikation

Zugriffsnummer NP_031668.3
Molekulargewicht 16.9 kDa
Quelle Mouse CD28, His Tag (CD8-M52H6) is expressed from human 293 cells (HEK293). It contains AA Asn 20 - Lys 149 (Accession # P31041-1).
Immunogen Asn 20 - Lys 149
Lagerungsbedingungen -20°C
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15826317
Unterlagen ansehen Aktionsdetails
ACROBiosystems
CD8-M52H6-1MG
1 mg
2626.22 CHF
1 Milligramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 10-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15816317
Unterlagen ansehen Aktionsdetails
ACROBiosystems
CD8-M52H6-100UG
100 μg
387.68 CHF
100 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 10-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

T-cell-specific surface glycoprotein CD28 is also known as TP44, is a single-pass type I membrane protein which contains one Ig-like V-type (immunoglobulin-like) domain. is one of the molecules expressed on T cells that provide co-stimulatory signals, which are required for T cell activation. CD28 is the receptor for CD80 (B7.1) and CD86 (B7.2). When activated by Toll-like receptor ligands, the CD80 expression is upregulated in antigen presenting cells (APCs). The CD86 expression on antigen presenting cells is constitutive. CD28 is the only B7 receptor constitutively expressed on naive T cells.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.9 kDa. The protein migrates as 30-35 kDa and 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

NP_031668.3
Mouse CD28, His Tag (CD8-M52H6) is expressed from human 293 cells (HEK293). It contains AA Asn 20 - Lys 149 (Accession # P31041-1).
-20°C
CD28
Mouse
PBS, pH7.4
HEK293
Immune Checkpoint Proteins
16.9 kDa
Asn 20 - Lys 149
CD28,Tp44
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications