Learn More
MAGE-1, Mouse, Clone: MA454, Abnova™
Mouse monoclonal antibody raised against MAGEA1.
Marke: Abnova MAB11321.100ug
Beschreibung
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. [provided by RefSeq
Spezifikation
MAGE-1 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
MAGEA1 | |
MAGEA1 | |
A recombinant protein fragment specific to MAGE-1. | |
100 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
MA454 | |
Mouse monoclonal antibody raised against MAGEA1. | |
In PBS (0.05% BSA, 0.05% sodium azide) | |
MAGE1/MGC9326 | |
Mouse | |
Protein G purification | |
RUO | |
4100 | |
Store at 4°C. | |
IgG1 κ |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.