missing translation for 'onlineSavingsMsg'
Learn More

CD5 Mouse anti-Human, APC-R700, Clone: UCHT2, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 565121

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15834099

  • 510.00 CHF / 1 Stück


Verfügbar ab
25-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The UCHT2 monoclonal antibody specifically binds to CD5. CD5 is a 67 kDa single-chain, type 1 transmembrane glycoprotein expressed on most thymocytes, the majority of peripheral T lymphocytes and a subpopulation of B cells. CD72 has been shown to be the natural ligand for CD5. CD5+ B cells produce polyreactive antibodies (mostly IgM).

This antibody was conjugated to BD Horizon APC-R700, which has been developed exclusively by BD Biosciences as a better alternative to Alexa Fluor™ 700. APC-R700 excites and emits at similar wavelengths to Alexa Fluor™ 700 yet exhibits significantly improved brightness. This dye can be excited by the red laser and detected with the same filter set as Alexa Fluor™ (eg, 730/45-nm filter).

Spezifikation

Spezifikation

CD5
UCHT2
CD5 antigen (p56-62); T1; Tp67; LEU1; Lymphocyte antigen T1/Leu-1
Affinity Purified
Human T cells
Affinity Chromatography
RUO
Primary
Human
Flow Cytometry
APC-R700
Aqueous buffered solution containing BSA, protein stabilizer, glycerol, and ≤0.09% sodium azide.
Murine
IgG1, κ
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only.