Learn More
CD171 Mouse anti-Human, BUV395, Clone: 5G3, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 565737
Additional Details : Gewicht : 0.01000kg
Beschreibung
The antibody was conjugated to BD Horizon BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348nm and an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.
Spezifikation
CD171 | |
Monoclonal | |
Brilliant Ultraviolet 395 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human SK-N-AS Neuroblastoma Cell Line | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
5G3 | |
L1 Neurite Cell Adhesion Molecule; N-CAM-L1; L1CAM ; CAML1; MIC5 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2a |
For Research Use Only.