missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELISA-Kit für Affen-IGF-2

Artikelnummer. 17738183
Ansicht ändern
Click to view available options
Menge:
10 x 96 Tests
Packungsgröße:
10 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17738183 10 x 96 Tests 10 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17738183 Lieferant Invitrogen™ Lieferanten-Nr. EP9RBX10
  • ONLINE EXCLUSIVE PRICE
    7200.00 CHF
    6984.00 CHF / 10 Stück
    Sparen 216.00 CHF
    3% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 04-09-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

ELISA

Monkey IGF-2 quantitates monkey IGF-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey IGF-2.

The IGFs are mitogenic, polypeptide growth factors that stimulate the proliferation and survival of various cell types, including muscle, bone, and cartilage tissue in vitro. IGFs are predominantly produced by the liver, although a variety of tissues produce the IGFs at distinctive times. The IGFs belong to the Insulin gene family, which also contains insulin and relaxin. The IGFs are similar to insulin by structure and function, but have a much higher growth-promoting activity than insulin. IGF-II expression is influenced by placenta lactogen, while IGF-I expression is regulated by growth hormone. Both IGF-I and IGF-II signal through the tyrosine kinase type I receptor (IGF-IR), but IGF-II can also signal through the IGF-II/Mannose-6-phosphate receptor. Mature IGFs are generated by proteolytic processing of inactive precursor proteins, which contain N-terminal and C-terminal propeptide regions.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer Q9Y6W8
Assay-Bereich 0,2 bis 50 ng/ml
Assay-Empfindlichkeit: 0,2 ng/ml
Konjugat Biotin
Immunassay-Kit-Format Sandwich ELISA Kit
Produkttyp ELISA
Probentyp Plasma, Serum, Überstand
Zielspezies Monkey
Zur Verwendung mit (Geräte) Kolorimetrisches Mikrotiterplatten-Lesegerät
Gen-Alias AL033362; C11orf43; FLJ22066; FLJ44734; GRDF; H-IGF-2; IGF; Igf2; Igf-2; IGFII; Igf-II; Insulin like growth factor; insulin like growth factor 2; insulin-like growth factor 2; insulin-like growth factor 2 (somatomedin A); insulin-like growth factor II; insulin-like growth factor II (IGF-II); Insulin-like growth factor II (somatomedin A); Insulin-like growth factor II Ala-25 Del; insulin-like growth factor II isoform 2 variant 4 preprotein; insulin-like growth factor type 2; M6pr; Mpr; MSA; multiplication-stimulating activity; Multiplication-stimulating polypeptide; Peg2; PP1446; PP9974; preptin; RNIGF2; Somatomedin-A; T3M-11-derived growth factorAL033362; C11orf43; FLJ22066; FLJ44734; GRDF; H-IGF-2; IGF; Igf2; Igf-2; IGFII; Igf-II; Insulin like growth factor; insulin like growth factor 2; insulin-like growth factor 2; insulin-like growth factor 2 (somatomedin A); insulin-like growth factor II; insulin-like growth factor II (IGF-II); Insulin-like growth factor II (somatomedin A); Insulin-like growth factor II Ala-25 Del; insulin-like growth factor II isoform 2 variant 4 preprotein; insulin-like growth factor type 2; M6pr; Mpr; MSA; multiplication-stimulating activity; Multiplication-stimulating polypeptide; Peg2; PP1446; PP9974; preptin; RNIGF2; Somatomedin-A; T3M-11-derived growth factor;
Gen-ID (Entrez) 29851
Gensymbol ICOS
Interassay-VK < 12%
Intraassay-VK < 10%
Kitinhalt Vorbeschichtete 96-Well-Platte|Standard|Assay-Verdünnungskonzentrat|Biotinylierte Detektionsantikörper|SAV-HRP|Waschpuffer|Chromogen|Stopplösung|Klebeplatten-Abdeckungen, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Marker oder Farbstoff HRP
Menge 10 x 96 Tests
Kennzeichnung RUO
Probenvolumen Überstand, 50 μl; Plasma, 50 μl; Serum, 50 μl
Versandbedingung Wet or Dry Ice
Lagerungsbedingungen 2 bis 8 °C
Ziel IGF2
Prüfzeit 1 h 20 min
Gesamte Assay-Zeit 4 h 45 min
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.