Learn More
Invitrogen™ Molecular Probes™ TSA™ Kit 25, with HRP-streptavidin and Alexa Fluor™ 594 tyramide
Beschreibung
- Includes HRP-streptavidin and bright, red-fluorescent Alexa Fluor 594 tyramide, which is spectrally similar to Texas Red dye
Cell Analysis, Cellular Imaging, Fluorescence In Situ Hybridization, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Immunohistochemistry (IHC), In Situ Hybridization (ISH)
Bestellinformation
Versandbedingung: Raumtemperatur
Spezifikation
Spezifikation
| Inhalt und Lagerung | Store in freezer (-5° to -30°C) and protect from light. |
| Menge | 1 Kit |