Learn More
Invitrogen™ Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 568 Conjugate
Beschreibung
Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B, IA2B2, IAB3, and IB4). The A subunit prefers N-acetyl-D-galactosamine end groups while the B subunit is selective for terminal a-D-galactosyl residues. The red fluorescent Alexa Fluor 568 isolectin GS-IB4 conjugate can be used in fluorescence microscopy.
Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Microbial Tracking
Bestellinformation
Shipping Condition: Room Temperature
Spezifikation
Spezifikation
| Menge | 500 μg |
| Quelle | Africa Shrub legume |
| Konjugat | Alexa Fluor 568 |
| Inhalt und Lagerung | Store in freezer (-5 to -30°C) and protect from light. |
| Markertyp | Alexa Fluor Dyes |
| Versandbedingung | Room Temperature |
| Form | Lyophilized |
| Protein-Subtyp | Lectins |
| Produktlinie | Alexa Fluor |
For Research Use Only. Not for use in diagnostic procedures.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.