Learn More
Invitrogen™ Molecular Probes™ aha-dCTP (5-Aminohexylacrylamido-dCTP)
Artikelnummer. 10053432
Click to view available options
Menge:
500 μl
Packungsgröße:
1 Stück
Artikelnummer. 10053432
Marke: Invitrogen™ A32768
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen
Supports two-step technique for improved DNA labeling results
Aha-dCTP (5-aminohexylacrylamido-dCTP) is modified at the C-5 position of cytosine with a unique hexylacrylamide moiety. aha-dCTP can be used to produce amine-modified DNA by conventional enzymatic incorporation methods such as reverse transcription. The amine-modified DNA can then be labeled with any amine-reactive dye to generate labeled nucleic acid hybridization probes. This two-step labeling technique consistently results in a uniform and high degree of DNA labeling that is difficult to obtain by other methods. In addition to this 2mM solution in TE (500μL unit size), it is available as a 50mM solution in TE (50μL unit size, Cat. No. A32769).
DNA and RNA Purification and Analysis, DNA Labeling, Nucleic Acid Labeling and Oligo Synthesis
TRUSTED_SUSTAINABILITY
Spezifikation
Produkttyp Nucleotide, aha-dCTP (5-Aminohexylacrylamido-dCTP)
Zur Verwendung mit (Anwendung) DNA and RNA Purification and Analysis, DNA Labeling, Nucleic Acid Labeling and Oligo Synthesis
Menge 500 μl