Learn More
MNT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA566935
Additional Details : Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: SIETLLEAARF LEWQAQQQQR AREEQERLRL EQEREQEQKK ANSLARLAHT LPVEEPRMEA PPLPLSPPA Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.
The Myc/Max/Mad network comprises a group of transcription factors that co-interact to regulate gene-specific transcriptional activation or repression. This gene encodes a protein member of the Myc/Max/Mad network. This protein has a basic-Helix-Loop-Helix-zipper domain (bHLHzip) with which it binds the canonical DNA sequence CANNTG, known as the E box, following heterodimerization with Max proteins. This protein is likely a transcriptional repressor and an antagonist of Myc-dependent transcriptional activation and cell growth. This protein represses transcription by binding to DNA binding proteins at its N-terminal Sin3-interaction domain.Spezifikation
MNT | |
Polyclonal | |
Unconjugated | |
MNT | |
BHLHD3; Class D basic helix-loop-helix protein 3; MAD6; max binding protein; MAX network transcriptional repressor; max-binding protein MNT; Max-interacting protein; MNT; MNT, MAX dimerization protein; MXD6; Myc antagonist MNT; Protein ROX; Rox | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4335 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99583 | |
MNT | |
Recombinant Human MNT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only