missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MNF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546265
Additional Details : Gewicht : 0.00010kg
Beschreibung
Peptide sequence: DTSFSGLSLE EYKLILSTDT LEELKEIDKG MWKKLQEKFA PKGPEEDHKA Sequence homology: Human: 100%; Mouse: 79%; Rat: 79%
MNF1 Polyclonal antibody specifically detects MNF1 in Human samples. It is validated for Western Blot.Spezifikation
MNF1 | |
Polyclonal | |
Unconjugated | |
UQCC2 | |
2900010M23Rik; bA6B20.2; breast cancer-associated protein SGA-81M; C6orf125; Cbp6; cytochrome B protein synthesis 6 homolog; M19; mitochondrial nucleoid factor 1; Mitochondrial protein M19; MNF1; RGD1306917; ubiquinol-cytochrome c reductase complex assembly factor 2; ubiquinol-cytochrome-c reductase complex assembly factor 2; UQCC2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84300 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BRT2 | |
UQCC2 | |
Synthetic peptide directed towards the C-terminal of human MNF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |