missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MMGT1 Monoclonal Antibody (OTI4D7)
GREENER_CHOICE

Artikelnummer. 15966984
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15966984 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Product Code. 15966984 Supplier Invitrogen™ Supplier No. MA527396

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMGT1 Monoclonal Antibody for Western Blot, IHC (P)

Mmgt1 encodes a protein that that mediates Mg(2+) transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMGT1
Anwendungen Immunohistochemistry (Paraffin), Western Blot
Klassifikation Monoclonal
Klon OTI4D7
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gen MMGT1
Gen-Zugriffsnummer Q8N4V1
Gen-Alias 9630048L06Rik; BC032271; EMBL:AAI68927.1}; EMC5; ER membrane protein complex subunit 5; Membrane magnesium transporter 1; membrane magnesium transporter 1 {ECO:0000250; Mir5116; mir-5116; Mmgt1; mmgt1 {ECO:0000250; RGD1566339; Tmem32; transmembrane protein 32; transmembrane protein 32 {ECO:0000312; UniProtKB:Q8K273}
Gensymbole MMGT1
Wirtsspezies Mouse
Immunogen Full length human recombinant protein of MMGT1 produced in E.coli.
Reinigungsverfahren Affinity Chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 93380
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.